Anti-SLITRK5 Mouse Monoclonal Antibody [clone: 310516]

Supplier: R & D Systems
MAB2587
220019-960EA 561.11 USD
220019-960
Anti-SLITRK5 Mouse Monoclonal Antibody [clone: 310516]
Antibodies
Type: Primary
Antigen: SLITRK5
Clonality: Monoclonal
Clone: 310516
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR