Anti-CSPG4 Mouse Monoclonal Antibody [clone: 7.1]

Supplier: R & D Systems
MAB25851
220019-959EA 577.02 USD
220019-959
Anti-CSPG4 Mouse Monoclonal Antibody [clone: 7.1]
Antibodies
Type: Primary
Antigen: CSPG4
Clonality: Monoclonal
Clone: 7.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR