Anti-CSPG4 Mouse Monoclonal Antibody [clone: LHM-2]

Supplier: R & D Systems
MAB2585
220019-958EA 639.2 USD
220019-958
Anti-CSPG4 Mouse Monoclonal Antibody [clone: LHM-2]
Antibodies
Type: Primary
Antigen: CSPG4
Clonality: Monoclonal
Clone: LHM-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR