Anti-CD300LB Rat Monoclonal Antibody [clone: 339003]

Supplier: R & D Systems
MAB2580
220019-957EA 633.42 USD
220019-957
Anti-CD300LB Rat Monoclonal Antibody [clone: 339003]
Antibodies
Type: Primary
Antigen: CD300LB
Clonality: Monoclonal
Clone: 339003
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR