Anti-CHODL Mouse Monoclonal Antibody [clone: 396305]

Supplier: R & D Systems
MAB2576
220019-956EA 577.02 USD
220019-956
Anti-CHODL Mouse Monoclonal Antibody [clone: 396305]
Antibodies
Type: Primary
Antigen: CHODL
Clonality: Monoclonal
Clone: 396305
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR