Anti-CRISP2 Mouse Monoclonal Antibody [clone: 350814]

Supplier: R & D Systems
MAB2575
220019-955EA 577.02 USD
220019-955
Anti-CRISP2 Mouse Monoclonal Antibody [clone: 350814]
Antibodies
Type: Primary
Antigen: CRISP2
Clonality: Monoclonal
Clone: 350814
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR