Anti-LILRA3 Mouse Monoclonal Antibody [clone: 293106]

Supplier: R & D Systems
MAB2574
220019-954EA 594.37 USD
220019-954
Anti-LILRA3 Mouse Monoclonal Antibody [clone: 293106]
Antibodies
Type: Primary
Antigen: LILRA3
Clonality: Monoclonal
Clone: 293106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR