Anti-SOX3 Mouse Monoclonal Antibody [clone: 287403]

Supplier: R & D Systems
MAB2569
220019-951EA 577.02 USD
220019-951
Anti-SOX3 Mouse Monoclonal Antibody [clone: 287403]
Antibodies
Type: Primary
Antigen: SOX3
Clonality: Monoclonal
Clone: 287403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR