Anti-STX2 Rat Monoclonal Antibody [clone: 338930]

Supplier: R & D Systems
MAB2568
220019-950EA 561.11 USD
220019-950
Anti-STX2 Rat Monoclonal Antibody [clone: 338930]
Antibodies
Type: Primary
Antigen: STX2
Clonality: Monoclonal
Clone: 338930
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR