Anti-DPPA3 Rat Monoclonal Antibody [clone: 283910]

Supplier: R & D Systems
MAB2566
220019-948EA 577.02 USD
220019-948
Anti-DPPA3 Rat Monoclonal Antibody [clone: 283910]
Antibodies
Type: Primary
Antigen: DPPA3
Clonality: Monoclonal
Clone: 283910
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR