Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 318311]

Supplier: R & D Systems
MAB2564
220019-947EA 600.15 USD
220019-947
Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 318311]
Antibodies
Type: Primary
Antigen: KIRREL2
Clonality: Monoclonal
Clone: 318311
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR