Anti-NGF Rat Monoclonal Antibody [clone: 347112]

Supplier: R & D Systems
MAB2562
220019-946EA 594.37 USD
220019-946
Anti-NGF Rat Monoclonal Antibody [clone: 347112]
Antibodies
Type: Primary
Antigen Symbol: NGF
Antigen Name: nerve growth factor (beta polypeptide)
Antigen Synonyms: Beta-NGF, HSAN5, NGFB
Clonality: Monoclonal
Clone: 347112
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR