Anti-TGFBI Rat Monoclonal Antibody [clone: 320305]

Supplier: R & D Systems
MAB2559
220019-943EA 577.02 USD
220019-943
Anti-TGFBI Rat Monoclonal Antibody [clone: 320305]
Antibodies
Type: Primary
Antigen: TGFBI
Clonality: Monoclonal
Clone: 320305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR