Anti-APCS Rat Monoclonal Antibody [clone: 313]

Supplier: R & D Systems
MAB2558
220019-942EA 577.02 USD
220019-942
Anti-APCS Rat Monoclonal Antibody [clone: 313]
Antibodies
Type: Primary
Antigen: APCS
Clonality: Monoclonal
Clone: 313
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR