Anti-GRN Rat Monoclonal Antibody [clone: 333708]

Supplier: R & D Systems
MAB2557
102885-478EA 772.25 USD
102885-478
Anti-GRN Rat Monoclonal Antibody [clone: 333708]
Antibodies
Type: Primary
Antigen Symbol: GRN
Antigen Name: granulin
Antigen Synonyms: Pgrn, epithelin
Clonality: Monoclonal
Clone: 333708
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR