Anti-MCFD2 Rat Monoclonal Antibody [clone: 300812]

Supplier: R & D Systems
MAB2556
220019-940EA 561.11 USD
220019-940
Anti-MCFD2 Rat Monoclonal Antibody [clone: 300812]
Antibodies
Type: Primary
Antigen: MCFD2
Clonality: Monoclonal
Clone: 300812
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR