Anti-CD200R1 Rat Monoclonal Antibody [clone: 330502]

Supplier: R & D Systems
MAB2554
220019-938EA 594.37 USD
220019-938
Anti-CD200R1 Rat Monoclonal Antibody [clone: 330502]
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Monoclonal
Clone: 330502
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR