Anti-GPNMB Mouse Monoclonal Antibody [clone: 303822]

Supplier: R & D Systems
MAB25501
220019-937EA 633.42 USD
220019-937
Anti-GPNMB Mouse Monoclonal Antibody [clone: 303822]
Antibodies
Type: Primary
Antigen Symbol: GPNMB
Antigen Name: glycoprotein (transmembrane) nmb
Antigen Synonyms: HGFIN, NMB
Clonality: Monoclonal
Clone: 303822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR