Anti-GPNMB Mouse Monoclonal Antibody [clone: 303802]

Supplier: R & D Systems
MAB2550
220019-936EA 594.37 USD
220019-936
Anti-GPNMB Mouse Monoclonal Antibody [clone: 303802]
Antibodies
Type: Primary
Antigen Symbol: GPNMB
Antigen Name: glycoprotein (transmembrane) nmb
Antigen Synonyms: HGFIN, NMB
Clonality: Monoclonal
Clone: 303802
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR