Anti-Laminin 1 Rat Monoclonal Antibody [clone: AL-3]

Supplier: R & D Systems
MAB25491
220019-935EA 577.02 USD
220019-935
Anti-Laminin 1 Rat Monoclonal Antibody [clone: AL-3]
Antibodies
Type: Primary
Antigen Symbol:
Antigen Name: Laminin-1
Antigen Synonyms:
Clonality: Monoclonal
Clone: AL-3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR