Anti-Laminin 1 Rat Monoclonal Antibody [clone: AL-2]

Supplier: R & D Systems
MAB2549
220019-934EA 633.42 USD
220019-934
Anti-Laminin 1 Rat Monoclonal Antibody [clone: AL-2]
Antibodies
Adhesion Blockade

Type: Primary
Antigen Symbol:
Antigen Name: Laminin-1
Antigen Synonyms:
Clonality: Monoclonal
Clone: AL-2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR