Anti-CTSF Rat Monoclonal Antibody [clone: 192204]

Supplier: R & D Systems
MAB2544
220019-929EA 763.57 USD
220019-929
Anti-CTSF Rat Monoclonal Antibody [clone: 192204]
Antibodies
Type: Primary
Antigen: CTSF
Clonality: Monoclonal
Clone: 192204
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR