Anti-CXCL5 Mouse Monoclonal Antibody [clone: 33103]

Supplier: R & D Systems
MAB2541
220019-928EA 763.57 USD
220019-928
Anti-CXCL5 Mouse Monoclonal Antibody [clone: 33103]
Antibodies
Type: Primary
Antigen: CXCL5
Clonality: Monoclonal
Clone: 33103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR