Anti-KLK15 Mouse Monoclonal Antibody [clone: 252819]

Supplier: R & D Systems
MAB2540
220019-927EA 763.57 USD
220019-927
Anti-KLK15 Mouse Monoclonal Antibody [clone: 252819]
Antibodies
Type: Primary
Antigen: KLK15
Clonality: Monoclonal
Clone: 252819
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR