Anti-DDR2 Mouse Monoclonal Antibody [clone: 290804]

Supplier: R & D Systems
MAB25381
220019-925EA 623.29 USD
220019-925
Anti-DDR2 Mouse Monoclonal Antibody [clone: 290804]
Antibodies
Type: Primary
Antigen: DDR2
Clonality: Monoclonal
Clone: 290804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR