Anti-DDR2 Mouse Monoclonal Antibody [clone: 290814]

Supplier: R & D Systems
MAB2538
220019-924EA 616.06 USD
220019-924
Anti-DDR2 Mouse Monoclonal Antibody [clone: 290814]
Antibodies
Type: Primary
Antigen: DDR2
Clonality: Monoclonal
Clone: 290814
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR