Anti-FGF21 Mouse Monoclonal Antibody [clone: 461804]

Supplier: R & D Systems
MAB25372
220019-923EA 594.37 USD
220019-923
Anti-FGF21 Mouse Monoclonal Antibody [clone: 461804]
Antibodies
Type: Primary
Antigen Symbol: FGF21
Antigen Name: fibroblast growth factor 21
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 461804
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR