Anti-FGF21 Mouse Monoclonal Antibody [clone: 315901]

Supplier: R & D Systems
MAB25371
220019-922EA 642.09 USD
220019-922
Anti-FGF21 Mouse Monoclonal Antibody [clone: 315901]
Antibodies
Type: Primary
Antigen Symbol: FGF21
Antigen Name: fibroblast growth factor 21
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 315901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR