Anti-MRC1 Rat Monoclonal Antibody [clone: 857615]

Supplier: R & D Systems
MAB25351
102885-476EA 594.37 USD
102885-476
Anti-MRC1 Rat Monoclonal Antibody [clone: 857615]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 857615
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR