Anti-MRC1 Rat Monoclonal Antibody [clone: 310301]

Supplier: R & D Systems
MAB2535
220019-919EA 594.37 USD
220019-919
Anti-MRC1 Rat Monoclonal Antibody [clone: 310301]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 310301
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR