Anti-MRC1 Mouse Monoclonal Antibody [clone: 685641]

Supplier: R & D Systems
MAB25342
102885-474EA 600.15 USD
102885-474
Anti-MRC1 Mouse Monoclonal Antibody [clone: 685641]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 685641
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR