Anti-MRC1 Mouse Monoclonal Antibody [clone: 685645]

Supplier: R & D Systems
MAB25341
102885-472EA 655.11 USD
102885-472
Anti-MRC1 Mouse Monoclonal Antibody [clone: 685645]
Antibodies
Simple Western

Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 685645
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR