Anti-MRC1 Rat Monoclonal Antibody [clone: 309210]

Supplier: R & D Systems
MAB2534
220019-918EA 594.37 USD
220019-918
Anti-MRC1 Rat Monoclonal Antibody [clone: 309210]
Antibodies
Type: Primary
Antigen: MRC1
Clonality: Monoclonal
Clone: 309210
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR