Anti-PVR Mouse Monoclonal Antibody [clone: 300907]

Supplier: R & D Systems
MAB25301
220019-917EA 655.11 USD
220019-917
Anti-PVR Mouse Monoclonal Antibody [clone: 300907]
Antibodies
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: 300907
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR