Anti-PVR Mouse Monoclonal Antibody [clone: 300903]

Supplier: R & D Systems
MAB2530
220019-916EA 594.37 USD
220019-916
Anti-PVR Mouse Monoclonal Antibody [clone: 300903]
Antibodies
Type: Primary
Antigen: PVR
Clonality: Monoclonal
Clone: 300903
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR