Anti-ADGRE5 Mouse Monoclonal Antibody [clone: 3895]

Supplier: R & D Systems
MAB25291
220019-914EA 763.57 USD
220019-914
Anti-ADGRE5 Mouse Monoclonal Antibody [clone: 3895]
Antibodies
Type: Primary
Antigen: ADGRE5
Clonality: Monoclonal
Clone: 3895
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR