Anti-ITGAV Mouse Monoclonal Antibody [clone: P5H9]

Supplier: R & D Systems
MAB2528
220019-912EA 623.29 USD
220019-912
Anti-ITGAV Mouse Monoclonal Antibody [clone: P5H9]
Antibodies
Adhesion Blockade

Type: Primary
Antigen: ITGAV
Clonality: Monoclonal
Clone: P5H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR