Anti-CHRDL2 Rat Monoclonal Antibody [clone: 328905]

Supplier: R & D Systems
MAB2520
220019-910EA 584.25 USD
220019-910
Anti-CHRDL2 Rat Monoclonal Antibody [clone: 328905]
Antibodies
Type: Primary
Antigen: CHRDL2
Clonality: Monoclonal
Clone: 328905
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR