Anti-CD36 Rat Monoclonal Antibody [clone: 324205]

Supplier: R & D Systems
MAB25191
102885-468EA 594.37 USD
102885-468
Anti-CD36 Rat Monoclonal Antibody [clone: 324205]
Antibodies
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 324205
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR