Anti-RPS6KA5 Mouse Monoclonal Antibody [clone: 252608]

Supplier: R & D Systems
MAB2518
220019-908EA 566.89 USD
220019-908
Anti-RPS6KA5 Mouse Monoclonal Antibody [clone: 252608]
Antibodies
Type: Primary
Antigen: RPS6KA5
Clonality: Monoclonal
Clone: 252608
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR