Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 275909]

Supplier: R & D Systems
MAB2515
220019-907EA 577.02 USD
220019-907
Anti-MAP2K3 Mouse Monoclonal Antibody [clone: 275909]
Antibodies
Type: Primary
Antigen: MAP2K3
Clonality: Monoclonal
Clone: 275909
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR