Anti-FGF7 Mouse Monoclonal Antibody [clone: 29568]

Supplier: R & D Systems
MAB2511
220019-905EA 772.25 USD
220019-905
Anti-FGF7 Mouse Monoclonal Antibody [clone: 29568]
Antibodies
Type: Primary
Antigen Symbol: FGF7
Antigen Name: fibroblast growth factor 7
Antigen Synonyms: HBGF-7, KGF
Clonality: Monoclonal
Clone: 29568
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR