Anti-APP Mouse Monoclonal Antibody [clone: 304904]

Supplier: R & D Systems
MAB2508
220019-903EA 577.02 USD
220019-903
Anti-APP Mouse Monoclonal Antibody [clone: 304904]
Antibodies
Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 304904
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR