Anti-GSK3B Mouse Monoclonal Antibody [clone: 609739]

Supplier: R & D Systems
MAB25062
102885-466EA 594.37 USD
102885-466
Anti-GSK3B Mouse Monoclonal Antibody [clone: 609739]
Antibodies
Type: Primary
Antigen Symbol: GSK3B
Antigen Name: glycogen synthase kinase 3 beta
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 609739
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR