Anti-GSK3B Rat Monoclonal Antibody [clone: 272535]

Supplier: R & D Systems
MAB25061
220019-902EA 577.02 USD
220019-902
Anti-GSK3B Rat Monoclonal Antibody [clone: 272535]
Antibodies
Type: Primary
Antigen Symbol: GSK3B
Antigen Name: glycogen synthase kinase 3 beta
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 272535
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR