Anti-GSK3B Rat Monoclonal Antibody [clone: 272536]

Supplier: R & D Systems
MAB2506
220019-901EA 594.37 USD
220019-901
Anti-GSK3B Rat Monoclonal Antibody [clone: 272536]
Antibodies
Type: Primary
Antigen Symbol: GSK3B
Antigen Name: glycogen synthase kinase 3 beta
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 272536
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR