Anti-CFTR Mouse Monoclonal Antibody [clone: 24-1]

Supplier: R & D Systems
MAB25031
220019-900EA 507.6 USD
220019-900
Anti-CFTR Mouse Monoclonal Antibody [clone: 24-1]
Antibodies
Type: Primary
Antigen: CFTR
Clonality: Monoclonal
Clone: 24-1
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR