Anti-LIF Mouse Monoclonal Antibody [clone: 9808]

Supplier: R & D Systems
MAB2501
220019-899EA 772.25 USD
220019-899
Anti-LIF Mouse Monoclonal Antibody [clone: 9808]
Antibodies
Type: Primary
Antigen: LIF
Clonality: Monoclonal
Clone: 9808
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR