Anti-ERAP1 Rat Monoclonal Antibody [clone: 325908]

Supplier: R & D Systems
MAB2500
220019-898EA 577.02 USD
220019-898
Anti-ERAP1 Rat Monoclonal Antibody [clone: 325908]
Antibodies
Type: Primary
Antigen: ERAP1
Clonality: Monoclonal
Clone: 325908
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR