Anti-KLKB1 Rat Monoclonal Antibody [clone: 337801]

Supplier: R & D Systems
MAB2498
220019-896EA 633.42 USD
220019-896
Anti-KLKB1 Rat Monoclonal Antibody [clone: 337801]
Antibodies
Type: Primary
Antigen: KLKB1
Clonality: Monoclonal
Clone: 337801
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR